Learn More
TARGETMOL CHEMICALS INC BARICITINIB PHOSPHATE 50MG
SDP
Catalog No. 502577146
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baricitinib phosphate (INCB028050) is a selective orally bioavailable JAK1/JAK2 inhibitor. purity: 99%
Catalog No. 50-257-7146 Supplier TARGETMOL CHEMICALS INC Supplier No. T236050MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baricitinib phosphate (INCB028050) is a selective orally bioavailable JAK1/JAK2 inhibitor. purity: 99%