Learn More
Medchemexpress LLC HY-15315A 50mg , Baricitinib (phosphate) CAS:1187595-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15315A50MG
Encompass_Preferred
HY-15315A 50mg Baricitinib (phosphate) CAS: 1187595-84-1 Baricitinib phosphate (LY3009104 phosphate; INCB028050 phosphate) is a selective orally bioavailable JAK1/JAK2 inhibitor with IC50 of 5.9 nM and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0903