Learn More
Selleck Chemical LLC Baricitinib phosphate
SDP
Catalog No. 5000003233
Encompass_Preferred
Baricitinib phosphate (INCB-028050 LY-3009104) is a selective JAK1 and JAK2 inhibitor with IC50 of 5 9 nM and 5 7 nM 70 and 10-fold selective versus JAK3 and Tyk2 no inhibition to c-Met and Chk2 Baricitinib is found to reduce or interrupt the passage of the virus into target cells and is used in the treatment research for COVID-19
Catalog No. 50-000-03233 Supplier Selleck Chemical LLC Supplier No. S57541G
Baricitinib phosphate (INCB-028050 LY-3009104) is a selective JAK1 and JAK2 inhibitor with IC50 of 5 9 nM and 5 7 nM 70 and 10-fold selective versus JAK3 and Tyk2 no inhibition to c-Met and Chk2 Baricitinib is found to reduce or interrupt the passage of the virus into target cells and is used in the treatment research for COVID-19