Learn More
Electron Microscopy Sciences Barrier Zipper Lock Small Pouch, 4¼" x 7½ "x 3"
SDP
Catalog No. NC1545649
Encompass_Preferred
These pouches are made of aluminum foil and multi-layered polyethylene. These are suitable to store items that have a sensitivity to oxygen and/or moisture. They have a tear-off notch above the zipper. Can be resealed by heat or the zipper. Also compatible with vacuum sealers.
Dimensions: 4¼" x 7½ "x 3"
Catalog No. NC1545649 Supplier Electron Microscopy Sciences Supplier No. 62313
These pouches are made of aluminum foil and multi-layered polyethylene. These are suitable to store items that have a sensitivity to oxygen and/or moisture. They have a tear-off notch above the zipper. Can be resealed by heat or the zipper. Also compatible with vacuum sealers.
Dimensions: 4¼" x 7½ "x 3"