Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Acetate, 99.0 to 102.0%, ACS Reagent, Solstice

Catalog No. 60045624 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-624 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-624 Supplier Solstice Supplier No. 243671100G

missing translation for 'validMainframeMsg'

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Color White
Quantity 100 g
Linear Formula (CH3COO)2Ba
UN Number NONH for all modes of transport
Beilstein 3693411
Formula Weight 255.42g/mol
Percent Purity 99.0 to 102.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder or crystals/granules
Density 2.46g/cm3
Chemical Name or Material Barium Acetate
Show More Show Less
Hazard Statement P261-P301 + P312 + P330
Precautionary Statement H302 + H332
EINECSNumber 208-849-0
RTECSNumber AF4550000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.