Learn More
Strem, An Ascensus Company CAS# 10553-31-8. 25g. Barium bromide, anhydrous, min. 95%. MFCD00003444
SDP
Catalog No. 5090115786
Encompass
CAS# 10553-31-8. 25g. Barium bromide, anhydrous, min. 95%. MFCD00003444. Molecular Weight: 297.16. Molecular Formula: BaBr2. Color/form: white pwdr. Strem# 93-5606. http://www.strem.com/catalog/v/93-5606/
Catalog No. 50-901-15786 Supplier Strem, An Ascensus Company Supplier No. 93560625G
CAS# 10553-31-8. 25g. Barium bromide, anhydrous, min. 95%. MFCD00003444. Molecular Weight: 297.16. Molecular Formula: BaBr2. Color/form: white pwdr. Strem# 93-5606. http://www.strem.com/catalog/v/93-5606/