Learn More
Strem, An Ascensus Company CAS# 513-77-9. 100g. Barium carbonate (99.9%-Ba, Sr < .03%). MFCD00003448
SDP
Catalog No. 5090112682
Encompass
CAS# 513-77-9. 100g. Barium carbonate (99.9%-Ba, Sr < .03%). MFCD00003448. Molecular Weight: 197.35. Molecular Formula: BaCO3. Color/form: -325 mesh pwdr. Strem# 56-1020. http://www.strem.com/catalog/v/56-1020/
Catalog No. 50-901-12682 Supplier Strem, An Ascensus Company Supplier No. 561020100G
CAS# 513-77-9. 100g. Barium carbonate (99.9%-Ba, Sr < .03%). MFCD00003448. Molecular Weight: 197.35. Molecular Formula: BaCO3. Color/form: -325 mesh pwdr. Strem# 56-1020. http://www.strem.com/catalog/v/56-1020/