Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC423440025
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423440025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423440025 Supplier Thermo Scientific Chemicals Supplier No. 423440025
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 513-77-9 | Molecular Formula: CBaO3 | Molecular Weight: 197.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point 1450.0°C
Melting Point 811.0°C
Color White
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula BaCO3
Merck Index 15, 965
Solubility Information Solubility in water: 0.002g/100mL (20°C)
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 4.43
Density 4.4300g/mL
Chemical Name or Material Barium carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-167-3
RTECSNumber CQ8600000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.