Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Carbonate, ACS, 99.0-101.0%, Spectrum™ Chemical
SDP

Catalog No. 18608968 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-968 125 g Poly Bottle
18-608-970 500 g Poly Bottle
18-608-972 2.5 kg Poly Bottle
18-608-971 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-968 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1015125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
IUPAC Name barium(2+) carbonate
SMILES [Ba++].[O-]C([O-])=O

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 125 g
Percent Purity 99 to 101%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Barium Carbonate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.