Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium carbonate, tech.

Catalog No. AA43478A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43478A3 2 kg
AA4347822 100 g
AA4347836 500 g
AA43478A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA43478A3 Supplier Thermo Scientific Chemicals Supplier No. 043478A3
Add to Cart

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers | CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Color White
Physical Form Powder
Quantity 2 kg
Solubility Information Almost insoluble in water. Soluble in NH4Cl solution,dilute HCl,HNO3,acetic acid. Insoluble in most alkalies.Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Odor Odorless
Density 4.43
Chemical Name or Material Barium carbonate
Show More Show Less
Hazard Category H302
Hazard Statement Avoid contact with skin and eyes.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-167-3
RTECSNumber CQ8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.