Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium chlorate monohydrate, 98%, Thermo Scientific™

Catalog No. p-7080560
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10128A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA10128A9 Supplier Thermo Scientific Chemicals Supplier No. A10128A9

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-38-9
Molecular Formula BaCl2H2O7
Molecular Weight (g/mol) 322.236
MDL Number MFCD00149147
InChI Key CIADYQPMTWAHHH-UHFFFAOYSA-L
Synonym barium chlorate monohydrate, barium 2+ dichlorate hydrate, ba.2clo3.h2o, chloric acid, bariumsalt 2:1, barium 2+ hydrate dichlorate
PubChem CID 6096944
IUPAC Name barium(2+);dichlorate;hydrate
SMILES O.[O-]Cl(=O)=O.[O-]Cl(=O)=O.[Ba+2]

Specifications

Melting Point 414°C
Quantity 10 kg
UN Number UN1445
Formula Weight 322.26 (304.24 Anhydrous)
Percent Purity 98%
Density 3.18
Chemical Name or Material Barium chlorate monohydrate
Hazard Category H271-H302+H332
Hazard Statement GHS H Statement
H271-H301-H331
May cause fire or explosion
strong oxidizer.
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM CHLORATE, SOLID
EINECSNumber 236-760-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.