Learn More
Aqua Solutions BARIUM CHLORIDE ACS GRADE 500G
SDP
Catalog No. NC9480424
Encompass_Preferred
Barium Chloride Dihydrate Reagent A.C.S., Crystal; 100%; White odorless crystals; 500g; 963 deg.C; 1560 deg.C; CAS: 10326-27-9; Store in a cool dry place
Catalog No. NC9480424 Supplier Aqua Solutions Supplier No. B0360500G
Barium Chloride Dihydrate Reagent A.C.S., Crystal; 100%; White odorless crystals; 500g; 963 deg.C; 1560 deg.C; CAS: 10326-27-9; Store in a cool dry place