Learn More
Strem, An Ascensus Company CAS# 10361-37-2. 100g. Barium chloride, anhydrous, 98+%. MFCD00003445
SDP
Catalog No. 5090115816
Encompass
CAS# 10361-37-2. 100g. Barium chloride, anhydrous, 98+%. MFCD00003445. Molecular Weight: 208.24. Molecular Formula: BaCl2. Color/form: white pwdr. Strem# 93-5652. http://www.strem.com/catalog/v/93-5652/
Catalog No. 50-901-15816 Supplier Strem, An Ascensus Company Supplier No. 935652100G
CAS# 10361-37-2. 100g. Barium chloride, anhydrous, 98+%. MFCD00003445. Molecular Weight: 208.24. Molecular Formula: BaCl2. Color/form: white pwdr. Strem# 93-5652. http://www.strem.com/catalog/v/93-5652/