Learn More
Strem, An Ascensus Company CAS# 10326-27-9. 250g. Barium chloride dihydrate, 99% (ACS). MFCD00149154
SDP
Catalog No. 5090115788
Encompass
CAS# 10326-27-9. 250g. Barium chloride dihydrate, 99% (ACS). MFCD00149154. Molecular Weight: 208.24 (244.27). Molecular Formula: BaCl2∙2H2O. Color/form: white xtl. Strem# 93-5610. http://www.strem.com/catalog/v/93-5610/
Catalog No. 50-901-15788 Supplier Strem, An Ascensus Company Supplier No. 935610250G
CAS# 10326-27-9. 250g. Barium chloride dihydrate, 99% (ACS). MFCD00149154. Molecular Weight: 208.24 (244.27). Molecular Formula: BaCl2∙2H2O. Color/form: white xtl. Strem# 93-5610. http://www.strem.com/catalog/v/93-5610/