Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, T.S., BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002406 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-406 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-406 Supplier Avantor Supplier No. 590701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7732-18-5
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Quantity 500 mL
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Barium Chloride, T.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.