Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Barium diphenylamine-4-sulfonate | 6211-24-1 | MFCD00007497 | 98.1% | 362.95 | C12H11NO3S · 1/2Ba | 5 G
SDP

Catalog No. 5000208402
Encompass_Preferred

Barium diphenylamine-4-sulfonate is a barium salt of diphenylamine-4-sulfonic acid supplied as a white to off-white solid powder for laboratory research. It is used as a dosing agent in analytical testing and as an intermediate in pharmaceutical and chemical research. For research use only.

  • White to off-white solid powder
  • Cas number 6211-24-1
  • Molecular weight 362.95 (as listed by supplier)
  • Purity 98.1%
  • Available in 1 g, 5 g, 10 g, and 50 g packs
  • Intended for use as a dosing agent and synthetic intermediate
  • For research use only; not for human use

Catalog No. 50-002-08402 Supplier Medchemexpress LLC Supplier No. HYW0878925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barium diphenylamine-4-sulfonate is a barium salt of diphenylamine-4-sulfonic acid supplied as a white to off-white solid powder for laboratory research. It is used as a dosing agent in analytical testing and as an intermediate in pharmaceutical and chemical research. For research use only.

  • White to off-white solid powder
  • Cas number 6211-24-1
  • Molecular weight 362.95 (as listed by supplier)
  • Purity 98.1%
  • Available in 1 g, 5 g, 10 g, and 50 g packs
  • Intended for use as a dosing agent and synthetic intermediate
  • For research use only; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.