Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium diphenylamine-4-sulfonate, For Redox Titration, ≥95%, MilliporeSigma™ Supelco™

Catalog No. 111014921 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-4921 5 g Poly Bottle
11-101-4919 25 g Poly Bottle
11-101-4920 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4921 Supplier MilliporeSigma Supelco Supplier No. D41305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym 4-(Phenylamino)benzenesulfonic acid barium salt; Diphenylamine-4-sulfonic acid barium salt
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Physical Form Powder
Quantity 5 g
Linear Formula (C12H10NO3S)2 Ba
Formula Weight 633.88
Percent Purity ≥95%
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material Barium Diphenylamine-4-sulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.