Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Diphenylamine Sulfonate, Reagents

Catalog No. 01337470
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-470 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-470 Supplier Reagents Holdings Llc Supplier No. C21472505C2
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barium Diphenylamine Sulfonate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym Barium diphenylamine-4-sulfonate, N-Phenylsulfanilic Acid Barium Salt
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color White to Gray
Physical Form Solid
Quantity 5 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0001
Chemical Name or Material Barium Diphenylamine Sulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.