Learn More
Strem, An Ascensus Company CAS# 12230-71-6. 250g. Barium hydroxide octahydrate, 98+% (ACS). MFCD00149152
SDP
Catalog No. 5090115796
Encompass
CAS# 12230-71-6. 250g. Barium hydroxide octahydrate, 98+% (ACS). MFCD00149152. Molecular Weight: 171.34 (315.46). Molecular Formula: Ba(OH)2∙8H2O. Color/form: white xtl. Strem# 93-5617. http://www.strem.com/catalog/v/93-5617/
Catalog No. 50-901-15796 Supplier Strem, An Ascensus Company Supplier No. 935617250G
CAS# 12230-71-6. 250g. Barium hydroxide octahydrate, 98+% (ACS). MFCD00149152. Molecular Weight: 171.34 (315.46). Molecular Formula: Ba(OH)2∙8H2O. Color/form: white xtl. Strem# 93-5617. http://www.strem.com/catalog/v/93-5617/