Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide Octahydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. B46500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B46-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46-500

missing translation for 'validMainframeMsg'

Specifications

Melting Point 78°C
Color White
CAS 12230-71-6
pH 12.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Molecular Formula BaH18O10
Linear Formula Ba(OH)2·8H2O
MDL Number MFCD00149152
UN Number 2923
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
IUPAC Name barium(2+) octahydrate dihydroxide
Molecular Weight (g/mol) 315.46
PubChem CID 17749109
Formula Weight 315.46
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 2.180g/cm³
Chemical Name or Material Barium Hydroxide Octahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Danger
Precautionary Statement Emergency Overview

Causes burns by all exposure routes. Ingestion causes severe swelling, severe damage to the delicate tissue and danger of perforation: Product is a corrosive material. Use of gastric lavage or emesis is contraindicated. Possible perforation of stomach or esophagus should be investigated. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.