Learn More
Strem, An Ascensus Company CAS# 13718-50-8. 25g. Barium iodide, anhydrous, min. 97%. MFCD00003452
SDP
Catalog No. 5090115798
Encompass
CAS# 13718-50-8. 25g. Barium iodide, anhydrous, min. 97%. MFCD00003452. Molecular Weight: 391.15. Molecular Formula: BaI2. Color/form: off-white pwdr. Strem# 93-5619. http://www.strem.com/catalog/v/93-5619/
Catalog No. 50-901-15798 Supplier Strem, An Ascensus Company Supplier No. 93561925G
CAS# 13718-50-8. 25g. Barium iodide, anhydrous, min. 97%. MFCD00003452. Molecular Weight: 391.15. Molecular Formula: BaI2. Color/form: off-white pwdr. Strem# 93-5619. http://www.strem.com/catalog/v/93-5619/