Learn More
Barium nitrate, 99%
Description
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Moisture sensitive. Incompatible with metal,alloy powders, combustible materials, reducing agents, acids, and acid anhydrides.
Specifications
Specifications
| Melting Point | 592°C (decomposition) |
| Quantity | 2500 g |
| Linear Formula | Ba(NO3)2 |
| UN Number | UN1446 |
| Merck Index | 14,983 |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 261.35 |
| Percent Purity | 99% |
| Sensitivity | Hygroscopic |
| Density | 3.24 |
| Show More |
Safety and Handling
| Hazard Category | H272-H301-H319-H332 |
| Hazard Statement | GHS H Statement H272-H302-H332 May intensify fire oxidizer. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM NITRATE |
| EINECSNumber | 233-020-5 |
| RTECSNumber | CQ9625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.