Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Solstice

Catalog No. 6000371 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-71 5 kg
60-003-70 500 g
60-045-911 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-003-71 Supplier Solstice Supplier No. 311285KG
Only null left
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥99%,≥52% Ba basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate

Specifications

Quantity 5 kg
Linear Formula Ba(NO3)2
UN Number 1446
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
Formula Weight 261.34g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Barium Nitrate, ACS reagent
missing translation for 'healthHazard2' P210-P220-P221-P261-P305 + P351 + P338-P370 + P378
missing translation for 'healthHazard3' H272-H302 + H332-H319
EINECSNumber 233-020-5
RTECSNumber CQ9625000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.