Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Solstice

Catalog No. 60045911 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-911 6 x 500 g
60-003-70 500 g
60-003-71 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-911 Supplier Solstice Supplier No. 311286X500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99%,≥52% Ba basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate

Specifications

Physical Form Solid
Quantity 6 x 500 g
Linear Formula Ba(NO3)2
UN Number UN1446
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 261.34
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Barium Nitrate, ACS reagent
Hazard Statement P210-P220-P221-P261-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H302 + H332-H319
DOTInformation 6.1(5.1) / PGII
EINECSNumber 233-020-5
RTECSNumber CQ9625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.