Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Nitrate, Reagent Grade Crystals, Innovating Science™
SCI_ED

Catalog No. S000532 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000532 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: May intensify fire; oxidizer, harmful if swallowed, harmful if inhaled.
Catalog No. S000532 Supplier Aldon Corporation Supplier No. IS11024
Add to Cart
Add to Cart
Danger: May intensify fire; oxidizer, harmful if swallowed, harmful if inhaled.

Chemical Identifiers

CAS 10022-31-8
Molecular Weight (g/mol) 261.35 g/mol

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Percent Purity 100%
Grade Reagent
Packaging HDPE
Chemical Name or Material Barium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.