Learn More
Strem, An Ascensus Company Barium oxide (99.5%-Ba) 25g
SDP
Catalog No. 5090115814
Encompass
CAS# 1304-28-5. 25g. Barium oxide (99.5%-Ba). MFCD00003453. Molecular Weight: 153.33. Molecular Formula: BaO. Color/form: -100 mesh off-white to light-gray pwdr. Strem# 93-5651. http://www.strem.com/catalog/v/93-5651/
Catalog No. 50-901-15814 Supplier Strem, An Ascensus Company Supplier No. 93565125G
CAS# 1304-28-5. 25g. Barium oxide (99.5%-Ba). MFCD00003453. Molecular Weight: 153.33. Molecular Formula: BaO. Color/form: -100 mesh off-white to light-gray pwdr. Strem# 93-5651. http://www.strem.com/catalog/v/93-5651/