Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Barium Perchlorate

Catalog No. 241733100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-173-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 24-173-3100G Supplier Sigma Aldrich Supplier No. 241733100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Barium Perchlorate

Specifications

CAS 3976-69-0
Density 1.055 g/mL at 20°C
Boiling Point 56°C to 58°C (11 mmHg, lit.)
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CH2CO2CH3
Quantity 100 g
MDL Number MFCD00063289
Synonym Barium diperchlorate; Perchloric acid barium salt
Molecular Weight (g/mol) 118.13 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.