Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium perchlorate trihydrate, Reagent Grade

Catalog No. p-7048850
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1165630 250 g
AA11656A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1165630 Supplier Thermo Scientific Chemicals Supplier No. 01165630
Only null left
Add to Cart

CAS: 10294-39-0 | BaCl2H6O11 | 390.264 g/mol

Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-39-0
Molecular Formula BaCl2H6O11
Molecular Weight (g/mol) 390.264
MDL Number MFCD00149148
InChI Key NBHDMFAHVJWOHD-UHFFFAOYSA-L
Synonym barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate
PubChem CID 53249218
IUPAC Name barium(2+);diperchlorate;trihydrate
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Physical Form Crystalline
Quantity 250 g
UN Number UN1447
Merck Index 14,987
Solubility Information Soluble in water, methanol; Slightly soluble in ethanol, ethyl acetate, acetone; Insoluble in ether
Formula Weight 390.29 (336.24 Anhydrous)
Grade Reagent
Refractive Index 1.533
Density 2.74 g/mL
Chemical Name or Material Barium perchlorate trihydrate
Hazard Category H271-H302+H332
Hazard Statement GHS H Statement
H271-H302-H332
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM PERCHLORATE, SOLID
EINECSNumber 236-710-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.