Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Peroxide, Powder, 88%, Spectrum™ Chemical
SDP

Catalog No. 18609021 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-021 25 g Amber Glass Bottle
18-609-022 125 g Amber Glass Bottle
18-609-024 500 g Amber Glass Bottle
18-609-025 2.5 kg Amber Glass Bottle
18-609-023 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-021 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B105325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1304-29-6

Specifications

CAS Max % 1
Physical Form Fine Crystals or Crystalline Powder
Quantity 25 g
Percent Purity 88%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Barium Peroxide, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.