Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Barium Sulfate
SCI_ED

Catalog No. s25191a Shop All Fisherbrand Products
Change view
Click to view available options
:
500g; Lab Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S25191A 500g; Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25191A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Color White to Yellow
CAS 7727-43-7
Quantity 500 g
Assay Percent Range >99%
Molecular Formula BaO4S
Linear Formula BaSO4
MDL Number MFCD00003455
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES [Ba++].[O-]S([O-])(=O)=O
IUPAC Name barium(2+) sulfate
Molecular Weight (g/mol) 233.38
PubChem CID 24414
Formula Weight 233.4
Grade Laboratory
Packaging Lab Grade
Density 4.4g/mL
Chemical Name or Material Barium Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.