Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Solstice

Catalog No. 6001905 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-05 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-019-05 Supplier Solstice Supplier No. 114321KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, Ph. Eur., 97.5-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
IUPAC Name barium(2+);sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 1 kg
Linear Formula BaSO4
UN Number NONH for all modes of transport
Formula Weight 233.39g/mol
Percent Purity 97.5 to 100.5%
Grade puriss.
Chemical Name or Material Barium Sulfate, Meets analytical specification of Ph.Eur.BP
EINECSNumber 231-784-4
RTECSNumber CR0600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.