Learn More
Sigma Aldrich Fine Chemicals Biosciences Barium sulfate ReagentPlus100G
Catalog No. 502899386
Encompass
Barium sulphate is widely employed as an inorganic filler. Its crystals belong to the rhombic crystal system. The rhombic crystals transforms to monoclinic form at 1148C and eventually it decomposes to barium oxide sulfur dioxide and oxygen at temperatures above 1400C.
Catalog No. 50-289-9386 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 243353100G
Barium sulphate is widely employed as an inorganic filler. Its crystals belong to the rhombic crystal system. The rhombic crystals transforms to monoclinic form at 1148C and eventually it decomposes to barium oxide sulfur dioxide and oxygen at temperatures above 1400C.