Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium tartrate, 98%

Catalog No. AAA1960418
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1960418 50 g
AAA1960430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1960418 Supplier Thermo Scientific Chemicals Supplier No. A1960418
Add to Cart

CAS: 5908-81-6 | C4H4BaO6 | 285.397 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5908-81-6
Molecular Formula C4H4BaO6
Molecular Weight (g/mol) 285.397
MDL Number MFCD00054437
InChI Key HQYOWPDUMCBSLQ-UHFFFAOYSA-L
Synonym barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate
PubChem CID 6453753
IUPAC Name barium(2+);2,3-dihydroxybutanedioate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2]

Specifications

Quantity 50 g
UN Number UN1564
Beilstein 6120238
Solubility Information Very slightly soluble in water
Formula Weight 285.41
Percent Purity 98%
Odor Odorless
Density 2.98
Chemical Name or Material Barium tartrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 227-615-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.