Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Barium Titanate(Iv)

Catalog No. 208108500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-810-8500G 500 g
20-810-82KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 20-810-8500G Supplier Sigma Aldrich Supplier No. 208108500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Barium titanate(IV) possess piezoelectric, ferroelectric, dielectric and thermoelectric properties. It is a biocompatible ceramic and is therefore used generally in drug delivery. It has been used:As a precursor to synthesize barium titanate microfibres to be used as building blocks for self-assembled nanoelectronic devices.In multi-material nanoscale 3D printing for developing capacitors, inductors and resistors.

Specifications

CAS 12047-27-7
Quantity 500 g
Molecular Formula BaO3Ti
Linear Formula BaTiO3
MDL Number MFCD00003447
Synonym Titanium barium oxide
Molecular Weight (g/mol) 233.19 g/mol
Density 6.08 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.