Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Barium titanate(IV)

Catalog No. 745952100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-595-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-595-2100G Supplier Sigma Aldrich Supplier No. 745952100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Barium Titanate is an effective lead free piezoelectric material and has been studied as a component of polymer nanocomposites and as a probe for cell imaging.

Specifications

CAS 12047-27-7
Quantity 100 g
Molecular Formula BaO3Ti
Linear Formula BaTiO3
MDL Number MFCD00003447
Synonym Barium titanium oxide; Titanium barium oxide
Molecular Weight (g/mol) 233.19
Percent Purity 99.9% trace metals basis
Density 6.08 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.