Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Barium titanate(IV)

Catalog No. 338842500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-884-2500G 500 g
33-884-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-884-2500G Supplier Sigma Aldrich Supplier No. 338842500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Barium titanate(IV) (BaTiO3) is a ceramic material that can be synthesized by reacting titanium oxide (TiO2) and barium salts in the presence of oleic acid as a stabilizing agent. It has a cubic crystal structure, which allows it to have ferroelectric, thermoelectric and piezoelectric properties. It forms a film with intense photoluminescence, which can be used for a wide range of electronic applications.BaTiO3 can be used to produce a nanocomposite for the fabrication of flexible nanogenerators. It can also be used as a filler material that can enhance the dielectric property of the composite.

Specifications

CAS 12047-27-7
Quantity 500 g
Molecular Formula BaO3Ti
Linear Formula BaTiO3
MDL Number MFCD00003447
Synonym Titanium barium oxide
Molecular Weight (g/mol) 233.19
Percent Purity 99.5% trace metals basis
Density 6.08 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.