Learn More
Hach Company BariVer 4 Barium Reagent Powder Pillows, pk/100
Supplier:  Hach Company 1206499
Encompass
For barium determination by the BariVer 4 Turbidimetric Method (Hach method 8014). Range: up to 100 mg/L. Sample Size: 25 mL. Pack of 100 powder pillows.
Catalog No. 50-199-4848