Learn More
Carnabio Usa Inc BARK1(ADRBK1)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0111110
Encompass
BARK1(ADRBK1), Full-length, Wild type, Amino Acid 1-689, AAA58391.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1050