Learn More
Carnabio Usa Inc BARK2(ADRBK2)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0111210
Encompass
BARK2(ADRBK2), Full-length, Wild type, Amino Acid 1-688, CAB45657.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0691