Learn More
Medchemexpress LLC HY-N0758 5mg Medchemexpress, Barlerin CAS:57420-46-9 Purity:>98%
SDP
Catalog No. 501933702
Encompass_Preferred
Medchemexpress, HY-N0758 5mg Barlerin CAS:57420-46-9 Barlerin (8-O-Acetyl shanzhiside methyl ester) is an iridoid glucoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant in Xi-zang. Barlerin (8-O-Acetyl shanzhiside methyl ester) could inhibt NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3702 Supplier Medchemexpress LLC Supplier No. HYN07585MG
Medchemexpress, HY-N0758 5mg Barlerin CAS:57420-46-9 Barlerin (8-O-Acetyl shanzhiside methyl ester) is an iridoid glucoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant in Xi-zang. Barlerin (8-O-Acetyl shanzhiside methyl ester) could inhibt NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.