Learn More
Electron Microscopy Sciences Barnes Dropping Bottle, 6 per pack
SDP
Catalog No. 501899140
Encompass_Preferred
The lower portion of the rubber bulb forms a tightly fitted stopper. Wheaton "400" borosilicate glass (meets USP Type I standards). Average drop size is 46ul. Bottle measures: 35 mm dia. x 103 mm high; 30 ml capacity. The bottles can be autoclaved with either dry heat or chemicals. The bulb and the pipette can be autoclaved in gas only.
Catalog No. 50-189-9140 Supplier Electron Microscopy Sciences Supplier No. 6421501
The lower portion of the rubber bulb forms a tightly fitted stopper. Wheaton "400" borosilicate glass (meets USP Type I standards). Average drop size is 46ul. Bottle measures: 35 mm dia. x 103 mm high; 30 ml capacity. The bottles can be autoclaved with either dry heat or chemicals. The bulb and the pipette can be autoclaved in gas only.