Learn More
Stoelting™ Barnes Maze
SDP
Catalog No. 10000305 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
Includes:
False Box
Goal Box
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Includes
10-000-305 Mouse
10-000-607 Mouse False Box
10-000-608 Mouse Goal Box
10-000-328 Rat
10-000-609 Rat False Box
10-000-610 Rat Goal Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10-000-305 Supplier Stoelting™ Supplier No. 60170
Only null left
Add to Cart
Add to Cart
Request a Quote
Barnes Maze task is similar to the Radial Arm and Morris Water mazes but without dietary restrictions and with less physical stress.
  • Designed for testing spatial learning and memory.
  • Within the maze, animals are motivated to escape from the brightly lit, open platform to a small dark recessed chamber (escape box).
  • During testing, the animal learns the spatial location of the escape box.
  • Optional false escape boxes help to remove visual cues that might be observed from a distance or through an open hole.
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Includes