Learn More
Stoelting™ Barnes Maze
SDP
Catalog No. 10000610 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
Includes:
—
False Box
Goal Box
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Includes
10-000-610 Rat Goal Box
10-000-305 Mouse —
10-000-607 Mouse False Box
10-000-608 Mouse Goal Box
10-000-328 Rat —
10-000-609 Rat False Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10-000-610 Supplier Stoelting™ Supplier No. 60270G
Add to Cart
Add to Cart
Barnes Maze task is similar to the Radial Arm and Morris Water mazes but without dietary restrictions and with less physical stress.
  • Designed for testing spatial learning and memory.
  • Within the maze, animals are motivated to escape from the brightly lit, open platform to a small dark recessed chamber (escape box).
  • During testing, the animal learns the spatial location of the escape box.
  • Optional false escape boxes help to remove visual cues that might be observed from a distance or through an open hole.
Specifications
For Use With (Application) Neuroscience
Animal Rat
Includes Goal Box