Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Barnidipine-d5 | 2771469-19-1 | 99.0% | 496.57 | C27H24D5N3O6 | 1 MG
SDP

Catalog No. 5000241494
Encompass_Preferred

Barnidipine-d5 is a deuterium-labeled reference standard of barnidipine intended for use as a stable isotope internal standard in analytical and pharmacokinetic assays. It is supplied as a small-mass, high-purity material suitable for LC-MS/MS quantitation and method development.

  • Deuterium-labeled analog (d5) to enable accurate internal standardization.
  • High chemical purity (≈99.0%) for reliable analytical results.
  • Molecular weight 496.57, formula C27H24D5N3O6.
  • Suitable for LC-MS/MS method development and pharmacokinetic studies.
  • Store sealed at -20°C; in solvent, store at -80°C for long-term stability.

Catalog No. 50-002-41494 Supplier Medchemexpress LLC Supplier No. HY107322AS11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barnidipine-d5 is a deuterium-labeled reference standard of barnidipine intended for use as a stable isotope internal standard in analytical and pharmacokinetic assays. It is supplied as a small-mass, high-purity material suitable for LC-MS/MS quantitation and method development.

  • Deuterium-labeled analog (d5) to enable accurate internal standardization.
  • High chemical purity (≈99.0%) for reliable analytical results.
  • Molecular weight 496.57, formula C27H24D5N3O6.
  • Suitable for LC-MS/MS method development and pharmacokinetic studies.
  • Store sealed at -20°C; in solvent, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.