Learn More
Medchemexpress LLC Barnidipine-d5 5mg | 2771469-19-1 | 5MG
SDP
Catalog No. 5000240453
Encompass_Preferred
Barnidipine-d5 is the deuterium-labeled Barnidipine (HY-107322A) Barnidipine-d5 (Mepirodipine) is an L-type calcium antagonist (CaA) with high affinity for [3H initrendipine binding sites (Ki 0 21 nmol/l) has selective action against CaA receptors[1 Barnidipine-d5 (Mepirodipine) is an antihypertensive agent and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action[2
Catalog No. 50-002-40453 Supplier Medchemexpress LLC Supplier No. HY107322AS15MG
Barnidipine-d5 is the deuterium-labeled Barnidipine (HY-107322A) Barnidipine-d5 (Mepirodipine) is an L-type calcium antagonist (CaA) with high affinity for [3H initrendipine binding sites (Ki 0 21 nmol/l) has selective action against CaA receptors[1 Barnidipine-d5 (Mepirodipine) is an antihypertensive agent and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action[2