Learn More
Medchemexpress LLC Barnidipine-d5 hydrochloride | 00-00-0 | 98.0% | 533.03 | C27H25D5ClN3O6 | 1 MG
SDP
Catalog No. 5000240672
Encompass_Preferred
Barnidipine-d5 hydrochloride is the deuterium-labeled analog of barnidipine provided as the hydrochloride salt for use as a tracer or internal standard in quantitative analytical workflows. It is intended for research use in NMR, GC-MS, and LC-MS applications and is supplied in a small analytical pack size.
Catalog No. 50-002-40672 Supplier Medchemexpress LLC Supplier No. HY107322S1MG
Add to Cart
Add to Cart
Barnidipine-d5 hydrochloride is the deuterium-labeled analog of barnidipine provided as the hydrochloride salt for use as a tracer or internal standard in quantitative analytical workflows. It is intended for research use in NMR, GC-MS, and LC-MS applications and is supplied in a small analytical pack size.