Learn More
Medchemexpress LLC Barnidipine hydrochloride | 104757-53-1 | MFCD00915772 | 528.00 g·mol⁻¹ | C27H30ClN3O6 | 100 MG
SDP
Catalog No. 5000240433
Encompass_Preferred
Barnidipine hydrochloride is a dihydropyridine calcium-channel blocker supplied as the hydrochloride salt for laboratory research. It is an orally active antihypertensive agent used in pharmacological and biochemical studies to investigate L-type calcium channel modulation and vascular effects.
  • Dihydropyridine calcium-channel blocker useful for cardiovascular research.
  • Provided as the hydrochloride salt to improve solubility.
  • Reported molecular formula C27H30ClN3O6 and molecular weight 528.00 g·mol⁻¹.
  • Suitable for in vitro and in vivo pharmacology studies (research use only).
  • Supplied in laboratory quantities for controlled experimental dosing.
Catalog No. 50-002-40433 Supplier Medchemexpress LLC Supplier No. HY107322100MG
Barnidipine hydrochloride is a dihydropyridine calcium-channel blocker supplied as the hydrochloride salt for laboratory research. It is an orally active antihypertensive agent used in pharmacological and biochemical studies to investigate L-type calcium channel modulation and vascular effects.
  • Dihydropyridine calcium-channel blocker useful for cardiovascular research.
  • Provided as the hydrochloride salt to improve solubility.
  • Reported molecular formula C27H30ClN3O6 and molecular weight 528.00 g·mol⁻¹.
  • Suitable for in vitro and in vivo pharmacology studies (research use only).
  • Supplied in laboratory quantities for controlled experimental dosing.