Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Barnidipine hydrochloride | 104757-53-1 | MFCD00915772 | 98.8% | 528.00 | C27H30ClN3O6 | 1 ML
SDP

Catalog No. 5000241107
Encompass_Preferred

Barnidipine hydrochloride is an L-type calcium channel antagonist used as a research reagent to study calcium-dependent signaling and vascular pharmacology. It is referenced as an orally active antihypertensive agent in the literature and is supplied with solubility and storage guidance for in vitro and in vivo use.

  • L-type calcium channel antagonist used in pharmacology research.
  • High purity material suitable for biochemical assays.
  • Chemical formula C27H30ClN3O6; molecular weight 528.00.
  • Available as 10 mM solution in DMSO (1 mL) and as solid weights (mg scale).
  • Storage: sealed at 4°C; in solvent store -80°C (up to 6 months) or -20°C (up to 1 month).
  • Solubility: highly soluble in DMSO; aqueous formulations require co-solvents per supplier recommendations.

Catalog No. 50-002-41107 Supplier Medchemexpress LLC Supplier No. HY10732210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barnidipine hydrochloride is an L-type calcium channel antagonist used as a research reagent to study calcium-dependent signaling and vascular pharmacology. It is referenced as an orally active antihypertensive agent in the literature and is supplied with solubility and storage guidance for in vitro and in vivo use.

  • L-type calcium channel antagonist used in pharmacology research.
  • High purity material suitable for biochemical assays.
  • Chemical formula C27H30ClN3O6; molecular weight 528.00.
  • Available as 10 mM solution in DMSO (1 mL) and as solid weights (mg scale).
  • Storage: sealed at 4°C; in solvent store -80°C (up to 6 months) or -20°C (up to 1 month).
  • Solubility: highly soluble in DMSO; aqueous formulations require co-solvents per supplier recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.