Learn More
Cayman Chemical Barnidipine (hydrochloride), Purity: ≥98%, CAS: 104757-53-1, 100 mg
SDP
Catalog No. 502984863
Encompass_Preferred
A calcium channel blocker that selectively antagonizes cardiovascular calcium channels; demonstrates antihypertensive activity by reducing peripheral vascular resistance; decreases blood pressure in spontaneously hypertensive rats
Catalog No. 50-298-4863 Supplier Cayman Chemical Supplier No. 20448100 MG
A calcium channel blocker that selectively antagonizes cardiovascular calcium channels; demonstrates antihypertensive activity by reducing peripheral vascular resistance; decreases blood pressure in spontaneously hypertensive rats