Learn More
Cayman Chemical BarnIdIpInhydrochlorIde 10mg
SDP
Catalog No. 502983613
Encompass_Preferred
A calcium channel blocker that selectively antagonizes cardiovascular calcium channels; demonstrates antihypertensive activity by reducing peripheral vascular resistance; decreases blood pressure in spontaneously hypertensive rats
Catalog No. 50-298-3613 Supplier Cayman Chemical Supplier No. 2044810 MG
A calcium channel blocker that selectively antagonizes cardiovascular calcium channels; demonstrates antihypertensive activity by reducing peripheral vascular resistance; decreases blood pressure in spontaneously hypertensive rats